2-[2-(2-Phenoxyethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049806-500MG 049806-2.5G
101889-124EA 364.62 USD
101889-124 101889-126
2-[2-(2-Phenoxyethoxy)ethyl]piperidine hydrochloride
2-[2-(2-Phenoxyethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560208
CAS Number: 1220030-05-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR