4-[2-(2-Phenoxyethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049804-500MG 049804-2.5G
101889-116EA 364.62 USD
101889-116 101889-118
4-[2-(2-Phenoxyethoxy)ethyl]piperidine hydrochloride
4-[2-(2-Phenoxyethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560206
CAS Number: 1220031-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR