3-[(2-Fluoroethoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
049803-500MG
049803-2.5G
101889-112EA
364.62
USD
101889-112
101889-114
3-[(2-Fluoroethoxy)methyl]pyrrolidine hydrochloride
3-[(2-Fluoroethoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₇H₁₅ClFNO |
MDL Number:
MFCD13560205 CAS Number: 1220038-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...