4-[2-(2-Fluoroethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049800-500MG 049800-2.5G
101889-100EA 364.62 USD
101889-100 101889-102
4-[2-(2-Fluoroethoxy)ethyl]piperidine hydrochloride
4-[2-(2-Fluoroethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClFNO
MDL Number: MFCD13560202
CAS Number: 1219967-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR