3-[2-(2-Cyclohexylethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049797-500MG 049797-2.5G
101889-088EA 364.62 USD
101889-088 101889-090
3-[2-(2-Cyclohexylethoxy)ethyl]piperidine hydrochloride
3-[2-(2-Cyclohexylethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₃₀ClNO
MDL Number: MFCD13560199
CAS Number: 1219979-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR