3-[(Cyclopropylmethoxy)methyl]pyrrolidine hydrochloride
Supplier: Matrix Scientific
049795-500MG
049795-2.5G
101889-080EA
186.15
USD
101889-080
101889-082
3-[(Cyclopropylmethoxy)methyl]pyrrolidine hydrochloride
3-[(Cyclopropylmethoxy)methyl]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₈ClNO |
MDL Number:
MFCD13560197 CAS Number: 1220038-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...