2-[2-(Cyclopropylmethoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
049794-500MG
049794-2.5G
101889-076EA
364.62
USD
101889-076
101889-078
2-[2-(Cyclopropylmethoxy)ethyl]piperidine hydrochloride
2-[2-(Cyclopropylmethoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₂ClNO |
MDL Number:
MFCD13560196 CAS Number: 1220016-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...