3-[(4-Chloro-3-ethylphenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

049791-500MG 049791-2.5G
101889-064EA 364.62 USD
101889-064 101889-066
3-[(4-Chloro-3-ethylphenoxy)methyl]pyrrolidine hydrochloride
3-[(4-Chloro-3-ethylphenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560193
CAS Number: 1219949-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR