2-[2-(4-Chloro-3-ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049790-500MG 049790-2.5G
101889-060EA 364.62 USD
101889-060 101889-062
2-[2-(4-Chloro-3-ethylphenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Chloro-3-ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560192
CAS Number: 1220029-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR