4-[2-(4-Chloro-3-ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049788-500MG 049788-2.5G
101889-052EA 364.62 USD
101889-052 101889-054
4-[2-(4-Chloro-3-ethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Chloro-3-ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560190
CAS Number: 1220034-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR