trans-2-Nitrocinnamic acid
Supplier: Matrix Scientific
Synonyms:
(E)-3-(2-Nitrophenyl)acrylic acid
048758-25G
048758-100G
101885-094EA
36.92
USD
101885-094
101885-096
trans-2-Nitrocinnamic acid
trans-2-Nitrocinnamic acid
Hazard: Irritant
Formula:
C₉H₇NO₄ MW: 193.16 g/mol Melting Pt: 243…245 °C |
MDL Number:
MFCD00007189 CAS Number: 1013-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...