6-Amino-5-bromonicotinic acid ≥98%

Supplier: Matrix Scientific

048731-1G 048731-5G
101885-040EA 89.23 USD
101885-040 101885-042
6-Amino-5-bromonicotinic acid ≥98%
6-Amino-5-bromonicotinic acid

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.0200 g/mol
MDL Number: MFCD11109515
CAS Number: 180340-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR