Methyl-2-fluoro-5-(trifluoromethyl)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Mthyl 2-fluoro-5-(trifluoromethyl)benzoate, Methyl 2-fluoro-5-(trifluoromethyl)benzoate
048708-1G
048708-5G
101884-962EA
30.77
USD
101884-962
102663-734
Methyl-2-fluoro-5-(trifluoromethyl)benzenecarboxylate ≥95%
Methyl-2-fluoro-5-(trifluoromethyl)benzenecarboxylate
Hazard: Irritant
Formula:
C₉H₆F₄O₂ MW: 222.14 g/mol |
MDL Number:
MFCD06203706 CAS Number: 556112-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...