2,4-Dibromo-5-(trifluoromethyl)aniline ≥95%

Supplier: Matrix Scientific

048678-500MG 048678-1G 048678-5G
101884-838EA 246.15 USD
101884-838 101884-840 101884-842
2,4-Dibromo-5-(trifluoromethyl)aniline ≥95%
2,4-Dibromo-5-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₇H₄Br₂F₃N
CAS Number: 24115-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR