2-Bromo-4-chloro-5-(trifluoromethyl)aniline ≥95%
Supplier: Matrix Scientific
048659-1G
048659-5G
101884-764EA
135.38
USD
101884-764
101884-766
2-Bromo-4-chloro-5-(trifluoromethyl)aniline ≥95%
2-Bromo-4-chloro-5-(trifluoromethyl)aniline
Hazard: Irritant
Formula:
C₇H₄BrClF₃N MW: 274.4700 g/mol Boiling Pt: 285 °C Density: 1.79 Flash Pt: 126 °C |
MDL Number:
MFCD11520118 CAS Number: 193090-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...