1-(6-Chloro-2-methyl-3-pyridinyl)-1-ethanone ≥95%
Supplier: Matrix Scientific
048593-500MG
048593-1G
101884-510EA
476.92
USD
101884-510
101884-512
1-(6-Chloro-2-methyl-3-pyridinyl)-1-ethanone ≥95%
1-(6-Chloro-2-methyl-3-pyridinyl)-1-ethanone
Hazard: Irritant
Formula:
C₈H₈ClNO |
MDL Number:
MFCD02661714 CAS Number: 439111-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...