3-(Chloromethyl)-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

048592-500MG 048592-1G
101884-506EA 384.62 USD
101884-506 101884-508
3-(Chloromethyl)-2-methoxypyridine ≥95%
3-(Chloromethyl)-2-methoxypyridine

Hazard: Irritant

Formula: C₇H₈ClNO
MDL Number: MFCD09880284
CAS Number: 162046-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR