2-Bromo-5-nitrobenzylalkohol ≥95%

Supplier: Matrix Scientific

048581-500MG 048581-1G
101884-458EA 538.46 USD
101884-458 101884-460
2-Bromo-5-nitrobenzylalkohol ≥95%
2-Bromo-5-nitrobenzylalkohol

Hazard: Irritant

Formula: C₇H₆BrNO₃
MDL Number: MFCD12031825
CAS Number: 332883-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR