Methyl-2-(1,2,3,4-tetrahydro-1-isoquinolinyl)acetate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetate
048551-500MG
048551-1G
101884-340EA
556.92
USD
101884-340
101884-342
Methyl-2-(1,2,3,4-tetrahydro-1-isoquinolinyl)acetate ≥95%
Methyl-2-(1,2,3,4-tetrahydro-1-isoquinolinyl)acetate
Hazard: Irritant
Formula:
C₁₂H₁₅NO₂ MW: 205.26 g/mol |
MDL Number:
MFCD12827811 CAS Number: 91640-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...