2,4-Dichloro-3-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

048524-1G 048524-5G
101884-210EA 292.31 USD
101884-210 101884-212
2,4-Dichloro-3-(trifluoromethyl)pyridine ≥98%
2,4-Dichloro-3-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂Cl₂F₃N
MDL Number: MFCD12547046
CAS Number: 1186194-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR