4-Chloro-2-iodo-5-(trifluoromethyl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-chloro-4-iodobenzotrifluoride

048515-500MG 048515-1G 048515-5G
101884-168EA 353.85 USD
101884-168 101884-170 101884-172
4-Chloro-2-iodo-5-(trifluoromethyl)aniline ≥95%
4-Chloro-2-iodo-5-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₇H₄ClF₃IN
MW: 321.466 g/mol
MDL Number: MFCD12827773
CAS Number: 852569-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR