1-(6-Bromo-2-pyridinyl)-1-ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(6-Bromopyridin-2-yl)ethanol

048512-500MG 048512-1G
101884-154EA 278.46 USD
101884-154 101884-156
1-(6-Bromo-2-pyridinyl)-1-ethanol ≥95%
1-(6-Bromo-2-pyridinyl)-1-ethanol

Hazard: Irritant

Formula: C₇H₈BrNO
MW: 202.06 g/mol
MDL Number: MFCD12198456
CAS Number: 139163-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR