4-Amino-3-bromo-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-methoxypyridin-4-amine, 3-Bromo-2-methoxy-4-pyridinamine

048511-500MG 048511-1G 048511-5G
101884-148EA 387.69 USD
101884-148 101884-150 101884-152
4-Amino-3-bromo-2-methoxypyridine ≥95%
4-Amino-3-bromo-2-methoxypyridine

Hazard: Irritant

Formula: C₆H₇BrN₂O
MDL Number: MFCD11656225
CAS Number: 215364-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR