5-Acetyl-3-bromo-6-methyl-2(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

048496-500MG 048496-1G 048496-5G
101884-084EA 556.92 USD
101884-084 101884-086 101884-088
5-Acetyl-3-bromo-6-methyl-2(1H)-pyridinone ≥95%
5-Acetyl-3-bromo-6-methyl-2(1H)-pyridinone

Hazard: Irritant

Formula: C₈H₈BrNO₂
MDL Number: MFCD12827806
CAS Number: 136117-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR