4-Cyano-3,5-difluoropyridine ≥98%

Supplier: Matrix Scientific

048492-1G 048492-5G
101884-072EA 61.54 USD
101884-072 101884-074
4-Cyano-3,5-difluoropyridine ≥98%
4-Cyano-3,5-difluoropyridine

Hazard: Irritant-Harmful

Formula: C₆H₂F₂N₂
MDL Number: MFCD13176565
CAS Number: 1214377-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR