Potassium(morpholin-4-ylmethyl)trifluoroborate ≥97%

Supplier: Matrix Scientific

Synonyms: Potassium trifluoro(morpholinomethyl)borate

048488-1G 048488-5G
101884-056EA 83.08 USD
101884-056 101884-058
Potassium(morpholin-4-ylmethyl)trifluoroborate ≥97%
Potassium (morpholin-4-ylmethyl)trifluoroborate

Hazard: Irritant

Formula: C₅H₁₀BF₃KNO
MDL Number: MFCD10566512
CAS Number: 936329-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR