3,3,4,4-Tetrafluoro-1,5-hexadiene ≥98%

Supplier: Matrix Scientific

048325-1G 048325-5G
101883-728EA 63.08 USD
101883-728 101883-730
3,3,4,4-Tetrafluoro-1,5-hexadiene ≥98%
3,3,4,4-Tetrafluoro-1,5-hexadiene

Hazard: Irritant

Formula: C₆H₆F₄
MDL Number: MFCD00042378
CAS Number: 1763-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR