6-Bromo[1,8]naphthyridin-2(1H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-1,8-naphthyridin-2-ol

048320-250MG 048320-1G
101883-716EA 136.92 USD
101883-716 101883-718
6-Bromo[1,8]naphthyridin-2(1H)-one ≥95%
6-Bromo[1,8]naphthyridin-2(1H)-one

Hazard: Irritant

Formula: C₈H₅BrN₂O
MW: 225.05 g/mol
MDL Number: MFCD11226504
CAS Number: 72754-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR