3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole ≥97%
Supplier: Matrix Scientific
048317-250MG
048317-1G
101883-708EA
67.69
USD
101883-708
101883-710
3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole ≥97%
3-(Chloromethyl)-5-(3-methoxyphenyl)-1,2,4-oxadiazole
Hazard: Irritant
Formula:
C₁₀H₉ClN₂O₂ MW: 224.65 g/mol |
MDL Number:
MFCD00084966 CAS Number: 175205-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...