1-[2-(Methylsulfanyl)-4-pyrimidinyl]-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(2-(Methylthio)pyrimidin-4-yl)ethanone, 4-Acetyl-2-(methylsulphanyl)pyrimidine
048306-500MG
048306-1G
101883-688EA
84.62
USD
101883-688
101883-690
1-[2-(Methylsulfanyl)-4-pyrimidinyl]-1-ethanone ≥95%
1-[2-(Methylsulfanyl)-4-pyrimidinyl]-1-ethanone
Hazard: Irritant
Formula:
C₇H₈N₂OS MW: 168.22 g/mol |
MDL Number:
MFCD09263472 CAS Number: 496863-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...