4-(4-Methylpiperazino)-2-(4-methylpiperidino)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(4-Methylpiperazin-1-yl)-2-(4-methylpiperidin-1-yl)aniline

048304-500MG 048304-1G
101883-682EA 747.69 USD
101883-682 101883-684
4-(4-Methylpiperazino)-2-(4-methylpiperidino)aniline ≥95%
4-(4-Methylpiperazino)-2-(4-methylpiperidino)aniline

Hazard: Irritant

Formula: C₁₇H₂₈N₄
MW: 288.44 g/mol
MDL Number: MFCD12025950
CAS Number: 1160474-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR