Methyl-2-(4-amino-3-iodophenyl)-2-methylpropanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(4-amino-3-iodophenyl)-2-methylpropanoate

048301-500MG 048301-1G 048301-5G
101883-670EA 747.69 USD
101883-670 101883-672 101883-674
Methyl-2-(4-amino-3-iodophenyl)-2-methylpropanoate ≥95%
Methyl-2-(4-amino-3-iodophenyl)-2-methylpropanoate

Hazard: Irritant

Formula: C₁₁H₁₄INO₂
MW: 319.14 g/mol
MDL Number: MFCD12025947
CAS Number: 756813-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR