Methyl-3-bromopicolinate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-bromo-2-pyridinecarboxylate

048299-1G 048299-5G
101883-662EA 30.77 USD
101883-662 101883-664
Methyl-3-bromopicolinate ≥97%
Methyl 3-bromopicolinate

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
MDL Number: MFCD12025946
CAS Number: 53636-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR