2-[(tert-Butoxycarbonyl)amino]butanoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
Boc-DL-Abu-OH, 2-Aminobutanoic acid, N-BOC protected
048298-500MG
048298-1G
101883-658EA
172.31
USD
101883-658
101883-660
2-[(tert-Butoxycarbonyl)amino]butanoic acid ≥95%
2-[(tert-Butoxycarbonyl)amino]butanoic acid
Hazard: Irritant
Formula:
C₉H₁₇NO₄ MW: 203.24 g/mol |
MDL Number:
MFCD00270358 CAS Number: 77284-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...