2-[(4-Aminophenyl)sulfanyl]acetonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[(Aminophenyl)thio]acetonitrile

048296-500MG 048296-1G
101883-650EA 556.92 USD
101883-650 101883-652
2-[(4-Aminophenyl)sulfanyl]acetonitrile ≥95%
2-[(4-Aminophenyl)sulfanyl]acetonitrile

Hazard: Irritant

Formula: C₈H₈N₂S
MW: 164.23 g/mol
MDL Number: MFCD09543129
CAS Number: 83591-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR