2-[1,1'-Biphenyl]-4-yl-1-ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-([1,1'-Biphenyl]-4-yl)ethanol, 4-(2-Hydroxyethyl)biphenyl

048295-500MG 048295-1G
101883-646EA 172.31 USD
101883-646 101883-648
2-[1,1'-Biphenyl]-4-yl-1-ethanol ≥95%
2-[1,1'-Biphenyl]-4-yl-1-ethanol

Hazard: Irritant

Formula: C₁₄H₁₄O
MW: 198.27 g/mol
MDL Number: MFCD09926182
CAS Number: 37729-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR