(S)-(-)-Chlocyphos ≥99%

Supplier: Matrix Scientific

Synonyms: (4S)-4-(2-Chlorophenyl)-2-hydroxy-5,5-dimethyl-1,3,2-dioxaphosphinane 2-oxide, (4S)-(-)-4-(2-Chlorophenyl)-5,5-dimethyl-2-hydroxy-1,3,2-dioxaphosphinane 2-oxide

048294-500MG 048294-1G
101883-642EA 172.31 USD
101883-642 101883-644
(S)-(-)-Chlocyphos ≥99%
(S)-(-)-Chlocyphos

Hazard: Irritant

Formula: C₁₁H₁₄ClO₄P
MW: 276.66 g/mol
MDL Number: MFCD00055615
CAS Number: 98674-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR