(S)-(-)-Anicyphos ≥99%

Supplier: Matrix Scientific

Synonyms: (4S)-(-)-5,5-Dimethyl-2-hydroxy-4-(2-methoxyphenyl)-1,3,2-dioxaphosphinane 2-oxide

048290-500MG 048290-1G
101883-626EA 181.54 USD
101883-626 101883-628
(S)-(-)-Anicyphos ≥99%
(S)-(-)-Anicyphos

Hazard: Irritant

Formula: C₁₂H₁₇O₅P
MW: 272.24 g/mol
MDL Number: MFCD00190173
CAS Number: 98674-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR