(R)-(+)-Anicyphos ≥99%

Supplier: Matrix Scientific

Synonyms: (4R)-(+)-5,5-Dimethyl-2-hydroxy-4-(2-methoxyphenyl)-1,3,2-dioxaphosphinane 2-oxide

048289-500MG 048289-1G
101883-622EA 181.54 USD
101883-622 101883-624
(R)-(+)-Anicyphos ≥99%
(R)-(+)-Anicyphos

Hazard: Irritant

Formula: C₁₂H₁₇O₅P
MW: 272.24 g/mol
MDL Number: MFCD00190172
CAS Number: 98674-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR