4-(Aminomethylene)-1H-isochromene-1,3(4H)-dione ≥95%

Supplier: Matrix Scientific

048287-500MG 048287-1G
101883-616EA 1058.46 USD
101883-616 101883-618
4-(Aminomethylene)-1H-isochromene-1,3(4H)-dione ≥95%
4-(Aminomethylene)-1H-isochromene-1,3(4H)-dione

Hazard: Irritant

Formula: C₁₀H₇NO₃
MDL Number: MFCD11553081
CAS Number: 78364-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR