2-[2-(Benzyloxy)-2-oxoethyl]acrylic acid ≥95%
Supplier: Matrix Scientific
048286-500MG
048286-1G
048286-5G
101883-610EA
356.92
USD
101883-610
101883-612
101883-614
2-[2-(Benzyloxy)-2-oxoethyl]acrylic acid ≥95%
2-[2-(Benzyloxy)-2-oxoethyl]acrylic acid
Hazard: Irritant
Formula:
C₁₂H₁₂O₄ |
MDL Number:
MFCD11553080 CAS Number: 48162-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...