(2,3-Dimethoxyphenyl)(4-pyridinyl)methanol ≥95%
Supplier: Matrix Scientific
048284-500MG
048284-1G
101883-602EA
375.38
USD
101883-602
101883-604
(2,3-Dimethoxyphenyl)(4-pyridinyl)methanol ≥95%
(2,3-Dimethoxyphenyl)(4-pyridinyl)methanol
Hazard: Irritant
Formula:
C₁₄H₁₅NO₃ |
MDL Number:
MFCD04966948 CAS Number: 243640-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...