2,3-Dihydroimidazo[2,1-b][1,3]thiazole-6-carbaldehyde ≥95%

Supplier: Matrix Scientific

048283-500MG 048283-1G
101883-598EA 1109.23 USD
101883-598 101883-600
2,3-Dihydroimidazo[2,1-b][1,3]thiazole-6-carbaldehyde ≥95%
2,3-Dihydroimidazo[2,1-b][1,3]thiazole-6-carbaldehyde

Hazard: Irritant

Formula: C₆H₆N₂OS
MDL Number: MFCD11553079
CAS Number: 157459-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR