4-Bromo-2,6-dimethylbenzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2,6-dimethylbenzonitrile

048282-500MG 048282-1G
101883-594EA 213.85 USD
101883-594 101883-596
4-Bromo-2,6-dimethylbenzenecarbonitrile ≥95%
4-Bromo-2,6-dimethylbenzenecarbonitrile

Hazard: Irritant

Formula: C₉H₈BrN
MW: 210.08 g/mol
MDL Number: MFCD01463960
CAS Number: 5757-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR