5,6-Dimethoxy-1-benzothiophene-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5,6-Dimethoxybenzo[b]thiophene-2-carboxylic acid
048280-500MG
048280-1G
101883-586EA
556.92
USD
101883-586
101883-588
5,6-Dimethoxy-1-benzothiophene-2-carboxylic acid ≥95%
5,6-Dimethoxy-1-benzothiophene-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₀O₄S MW: 238.26 g/mol |
MDL Number:
MFCD08275132 CAS Number: 23046-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...