N'-Hydroxy-5,6-dimethoxy-1-benzothiophene-2-carboximidamide hydrochloride ≥95%

Supplier: Matrix Scientific

048279-500MG 048279-1G
101883-582EA 556.92 USD
101883-582 101883-584
N'-Hydroxy-5,6-dimethoxy-1-benzothiophene-2-carboximidamide hydrochloride ≥95%
N'-Hydroxy-5,6-dimethoxy-1-benzothiophene-2-carboximidamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₃S
MDL Number: MFCD12922723
CAS Number: 101041-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR