Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic acid ≥95%
Supplier: Matrix Scientific
048278-500MG
048278-1G
101883-578EA
556.92
USD
101883-578
101883-580
Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic acid ≥95%
Bicyclo[2.2.2]oct-5-ene-2,3-dicarboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₂O₄ |
MDL Number:
MFCD00267156 CAS Number: 28871-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...