2-[1-(tert-Butoxycarbonyl)-4-piperidinyl]-1,3-thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

048277-500MG 048277-1G
101883-574EA 375.38 USD
101883-574 101883-576
2-[1-(tert-Butoxycarbonyl)-4-piperidinyl]-1,3-thiazole-4-carboxylic acid ≥95%
2-[1-(tert-Butoxycarbonyl)-4-piperidinyl]-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₄S
MDL Number: MFCD06658980
CAS Number: 365413-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR