2,4-Dihydroxy-5-methoxypyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methoxy-2,4-pyrimidinediol

048276-1G 048276-5G
101883-572EA 15.38 USD
101883-572 102551-384
2,4-Dihydroxy-5-methoxypyrimidine ≥95%
2,4-Dihydroxy-5-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₆N₂O₃
MW: 142.1100 g/mol
MDL Number: MFCD00187903
CAS Number: 6623-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR