DL-O-Methylserine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-3-methoxypropanoic acid
048271-1G
048271-5G
101883-556EA
198.46
USD
101883-556
101883-558
DL-O-Methylserine ≥95%
DL-O-Methylserine
Hazard: Irritant
Formula:
C₄H₉NO₃ MW: 119.12 g/mol |
MDL Number:
MFCD00037773 CAS Number: 19794-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...